Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM132A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546452
Description
Peptide sequence: MRRWAWAAVV VLLGPQLVLL GGVGARREAQ RTQQPGQRAD PPNATASASS Sequence homology: Human: 100%; Zebrafish: 82%.
Insulin-sensitizing adipocyte-secreted protein (adipokine) that regulates glucose metabolism in liver and adipose tissue. Promotes glucose uptake in adipocytes and suppresses de novo glucose production in hepatocytes via the PI3K-Akt signaling pathway. Administration lead to reduction of blood glucose. Able to attenuate inflammation in fat tissue. [UniProt].
Specifications
FAM132A | |
Polyclonal | |
Unconjugated | |
C1QTNF12 | |
Adipolin; Adipolin cleaved form; Adipolin fC1QTNF12; Adipolin fCTRP12; Adipolin full-length form; Adipolin gC1QTNF12; Adipolin gCTRP12; Adipose-derived insulin-sensitizing factor; C1q and TNF related 12; C1q and TNF related protein 12; C1q domain containing 2; C1q/TNF-related protein 12; C1QDC2; C1QTNF12; Complement C1q tumor necrosis factor-related protein 12; CTRP12; FAM132A; family with sequence similarity 132 member A; family with sequence similarity 132, member A; protein FAM132A; zgc:158358 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
388581 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5T7M4 | |
C1QTNF12 | |
Synthetic peptide directed towards the N-terminal of human FAM132A (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction