Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM132B Polyclonal Antibody, Invitrogen™

Catalog No. PIPA567448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567448 Supplier Thermo Scientific Supplier No. PA567448

Rabbit Polyclonal Antibody

Immunogen sequence: EPTAERAHSVD PRDAWMLFVR QSDKGVNGKK RSRGKAKKLK FGLPGP Highest antigen sequence identity to the following orthologs - mouse 74%, rat 79%.

Iron-regulatory hormone that acts as an erythroid regulator after hemorrhage: produced by erythroblasts following blood loss and mediates suppression of hepcidin (HAMP) expression in the liver, thereby promoting increased iron absorption and mobilization from stores. Promotes lipid uptake into adipocytes and hepatocytes via transcriptional up-regulation of genes involved in fatty acid uptake.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM132B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERFE
Gene Accession No. Q4G0M1
Gene Alias C1QTNF15; complement C1q tumor necrosis factor-related protein 15; CTRP15; ERFE; Erythroferrone; FAM132B; family with sequence similarity 132 member B; family with sequence similarity 132, member B; myonectin; protein FAM132B
Gene Symbols ERFE
Host Species Rabbit
Immunogen Recombinant Human FAM132B
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151176
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.