Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM133B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570027 Supplier Invitrogen™ Supplier No. PA570027
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 86%; Horse: 92%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 79%; Rat: 86%; Yeast: 79%; Zebrafish: 100%.

FAM133B (Family With Sequence Similarity 133 Member B) is a Protein Coding gene. An important paralog of this gene isFAM133A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM133B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM133B
Gene Accession No. Q9CVI2
Gene Alias 2900022K02Rik; 5830415L20Rik; FAM133; Fam133b; family with sequence similarity 133 member B; family with sequence similarity 133, member B; LOC553430 protein; protein FAM133; Protein FAM133B; RGD1306302; si:ch211-287b5.3; Unknown (protein for IMAGE:8130787); wu:fc17e01; zgc:153800
Gene Symbols FAM133B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse FAM133B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68152
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.