Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM134A Polyclonal Antibody

Catalog No. PIPA564940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564940 Supplier Invitrogen™ Supplier No. PA564940

Rabbit Polyclonal Antibody

Immunogen sequence: SGGFSVSRATT PQLTDVSEDL DQQSLPSEPE ETLSRDLGEG EEGELAPPED LLGRPQALSR QALDSEEEEE DVAAKETLLR LSSPLHFVNT HFNGAGSPPD GVKCSPGGPV ETLSPETVSG GLTALPGTLS PPLCLVGSDP A Highest antigen sequence identity to the following orthologs - mouse 77%, rat 80%.

FAM134A is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM134A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RETREG2
Gene Accession No. Q8NC44
Gene Alias C2H2orf17; C2orf17; Fam134a; family with sequence similarity 134 member A; family with sequence similarity 134, member A; MAG2; MAG-2; metastasis associated gene-2; protein FAM134A; reticulophagy regulator 2; reticulophagy regulator 2; protein FAM134A; reticulophagy regulator family member 2; Retreg2; RGD1306844; zgc:158472
Gene Symbols RETREG2
Host Species Rabbit
Immunogen Recombinant Human FAM134A.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.