Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM134B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100514
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100514 Supplier Invitrogen™ Supplier No. PA5100514
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FAM134B.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM134B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RETREG1
Gene Accession No. Q8VE91, Q9H6L5
Gene Alias 1810015C04Rik; AU015349; FAM134B; family with sequence similarity 134 member B; family with sequence similarity 134, member B; hypothetical protein FLJ20152; hypothetical protein LOC540068; JK1; JK-1; protein FAM134B; Protein FAM134B-like protein; reticulophagy receptor 1; reticulophagy receptor FAM134B; reticulophagy regulator 1; Retreg1
Gene Symbols RETREG1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RETREG1(Accession Q9H6L5), corresponding to amino acid residues L301-P351.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54463, 66270
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.