Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM134B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542647 Supplier Invitrogen™ Supplier No. PA542647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LSVSDTDVSE VSWTDNGTFN LSEGYTPQTD TSDDLDRPSE EVFSRDLSDF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM134B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RETREG1
Gene Accession No. Q9H6L5
Gene Alias 1810015C04Rik; AU015349; FAM134B; family with sequence similarity 134 member B; family with sequence similarity 134, member B; hypothetical protein FLJ20152; hypothetical protein LOC540068; JK1; JK-1; protein FAM134B; Protein FAM134B-like protein; reticulophagy receptor 1; reticulophagy receptor FAM134B; reticulophagy regulator 1; Retreg1
Gene Symbols RETREG1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human FAM134B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54463
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.