Learn More
Invitrogen™ FAM134B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113800
Description
Immunogen sequence: DFGIGEYINQ KKRERSEADK EKSHKDDSEL DFSALCPKIS LTVAAKELSV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 77%.
The function remains unknown.
Specifications
| FAM134B | |
| Polyclonal | |
| Unconjugated | |
| RETREG1 | |
| 1810015C04Rik; AU015349; FAM134B; family with sequence similarity 134 member B; family with sequence similarity 134, member B; hypothetical protein FLJ20152; hypothetical protein LOC540068; JK1; JK-1; protein FAM134B; Protein FAM134B-like protein; reticulophagy receptor 1; reticulophagy receptor FAM134B; reticulophagy regulator 1; Retreg1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54463 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H6L5 | |
| RETREG1 | |
| Synthetic peptide directed towards the middle region of human FAM134B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.