Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM134B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982643
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2643 Supplier Bioss Supplier No. BS13136R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function remains unknown.

Specifications

Antigen FAM134B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RETREG1
Gene Accession No. Q9H6L5
Gene Alias 1810015C04Rik; AU015349; BOS_19918; FAM134B; family with sequence similarity 134 member B; family with sequence similarity 134, member B; JK1; JK-1; LOC100343570; protein FAM134B; Reticulophagy receptor 1; reticulophagy receptor FAM134B; Reticulophagy regulator 1; RETREG1; uncharacterized LOC100343570
Gene Symbols RETREG1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM134B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54463
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.