Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM134C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553433
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553433 Supplier Invitrogen™ Supplier No. PA553433
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DFPSINMDPA GLDDEDDTSI GMPSLMYRSP PGAEEPQAPP ASRDEAALPE LLLGALPVGS NLTSNLASLV SQGMIQLALS GASQPGPSGA PAQRATRGFL RSPSSDLDTD AEGDDFELLD QSE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.

RETREG3 gene ontology annotations related to this gene include positive regulation of neuron projection development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM134C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RETREG3
Gene Accession No. Q86VR2, Q9CQV4
Gene Alias 1300010M03Rik; 4933404C01Rik; AI551748; DKFZp686B1036; Fam134c; family with sequence similarity 134 member C; family with sequence similarity 134, member C; FLJ33806; protein FAM134C; Reticulophagy regulator 3; RETREG3; RGD1561189
Gene Symbols RETREG3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM134C. Recombinant protein control fragment (Product #RP-91078).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162427, 360632, 67998
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.