Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM135B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988790
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8790 Supplier Bioss Supplier No. BS9063R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM135B gene ontology annotations related to this gene include cellular lipid metabolic process.

Specifications

Antigen FAM135B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM135B
Gene Accession No. Q49AJ0
Gene Alias 1700010C24Rik; A830008O07Rik; BOS_13681; C8orfK32; FAM135B; family with sequence similarity 135 member B; family with sequence similarity 135, member B; LOC100359280; LOC100726490; Protein FAM135B; RGD1308133; uncharacterized LOC100359280
Gene Symbols FAM135B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM135B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51059
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.