Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM136A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556344
Description
Immunogen sequence: SELEKFQDRL ARCTMHCNDK AKDSIDAGSK ELQVKQQLDS CVTKCVDDHM HLIPTMTKKM KEALLSIG Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
This gene encodes a mitochondrially localized protein that is highly conserved across species. The gene is expressed in a variety of tissues including human lymphoblast cells and rat neurosensorial epithelium of the cristaampullaris. A mutation in this gene has been associated with familial Meniere's disease, a chronic disorder of the inner ear. Several pseudogenes of this gene are found on other chromosomes. Alternative splicing results in multiple transcript variants.
Specifications
| FAM136A | |
| Polyclonal | |
| Unconjugated | |
| FAM136A | |
| FAM136A; family with sequence similarity 136 member A; family with sequence similarity 136, member A; protein FAM136A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84908 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96C01 | |
| FAM136A | |
| Recombinant protein corresponding to Human FAM136A. Recombinant protein control fragment (Product #RP-95962). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction