Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM136A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556345 Supplier Invitrogen™ Supplier No. PA556345
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAELQQLRVQ EAVESMVKSL ERENIRKMQG LMFRCSASCC EDSQASMKQV HQCIERCHVP LAQAQALVT Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

This gene encodes a mitochondrially localized protein that is highly conserved across species. The gene is expressed in a variety of tissues including human lymphoblast cells and rat neurosensorial epithelium of the cristaampullaris. A mutation in this gene has been associated with familial Meniere's disease, a chronic disorder of the inner ear. Several pseudogenes of this gene are found on other chromosomes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM136A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM136A
Gene Accession No. Q96C01
Gene Alias FAM136A; family with sequence similarity 136 member A; family with sequence similarity 136, member A; protein FAM136A
Gene Symbols FAM136A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM136A. Recombinant protein control fragment (Product #RP-95961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84908
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.