Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM13A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100515
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100515 Supplier Invitrogen™ Supplier No. PA5100515
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FAM13A.

FAM13A, also named as FAM13A1 and KIAA0914, belongs to the FAM13 family. It is associated with chronic obstructive pulmonary disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM13A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FAM13A
Gene Accession No. O94988
Gene Alias AI462012; ARHGAP48; D430015B01Rik; Fam13a; Fam13a1; FAM13A1_v2 protein; family with sequence similarity 13 member A; family with sequence similarity 13, member A; family with sequence similarity 13, member A1; KIAA0914; Precm1; Precm1 protein; Protein FAM13A; RGD1309807
Gene Symbols FAM13A
Host Species Rabbit
Immunogen A synthesized peptide derived from human FAM13A(Accession O94988), corresponding to amino acid residues K253-D303.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10144
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.