Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM13B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565314
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565314 Supplier Invitrogen™ Supplier No. PA565314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPSLSNCNSVL ANKIFGIPLD ELQQGGHPDN EVPFIVRHVV DYIEEHGGLE QQGLFQVNGN AETVEWLRQR YDSGEEVDLV KEADVPSAI Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

FAM13B is a 915 amino acid protein that is encoded by a gene that maps to human chromosome 5. With 181 million base pairs encoding around 1,000 genes, chromosome 5 is about 6% of human genomic DNA. It is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM13B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM13B
Gene Accession No. Q9NYF5
Gene Alias 2610024E20Rik; ARHGAP49; AW060714; AW546153; C5orf5; Fam13b; FAM13B1; family with sequence similarity 13 member B; family with sequence similarity 13, member B; family with sequence similarity 13, member B1; GAP-like protein N61; KHCHP; N61; protein FAM13B; RGD1310484
Gene Symbols FAM13B
Host Species Rabbit
Immunogen Recombinant Human FAM13B. Recombinant protein control fragment (Product #RP-106016).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51306
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.