Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM13C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558152 Supplier Invitrogen™ Supplier No. PA558152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVHGVKDPAP ASTQSVLADG TDSADPSPVH KDGQNEADSA PEDLHSVGTS RLLYHITDGD NPLLSPRCSI FSQSQRFNLD PESAPSPPST Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

FAM13C1 belongs to the FAM13 family. The exact function of FAM13C1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM13C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM13C
Gene Accession No. Q8NE31
Gene Alias 1200015N20Rik; C030038O19Rik; Fam13c; FAM13C1; family with sequence similarity 13 member C; family with sequence similarity 13, member C; family with sequence similarity 13, member C1; mKIAA1796; protein FAM13C; RGD1310149
Gene Symbols FAM13C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM13C. Recombinant protein control fragment (Product #RP-97104).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220965
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.