Learn More
Invitrogen™ FAM13C Polyclonal Antibody

Description
Positive Control: Human brain tissue lysates, mouse colon tissue, HT-29, SH-SY5Y. Subcellular Location: Nuclear and/or cytoplasmic expression in most tissues, some also show membranous expression.
Specifications
Specifications
| Antigen | FAM13C |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | FAM13C |
| Gene Accession No. | Q8NE31, Q9DBR2 |
| Gene Alias | 1200015N20Rik; C030038O19Rik; Fam13c; FAM13C1; family with sequence similarity 13 member C; family with sequence similarity 13, member C; family with sequence similarity 13, member C1; mKIAA1796; protein FAM13C; RGD1310149 |
| Gene Symbols | FAM13C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.