Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM13C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544150
Description
Peptide sequence: TEHVVSSQSE CQVRAGTPAH ESPQNNAFKC QETVRLQPRI DQRTAISPKD Sequence homology: Dog: 92%; Guinea Pig: 85%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 92%.
FAM13C1 belongs to the FAM13 family. The exact function of FAM13C1 remains unknown.
Specifications
FAM13C | |
Polyclonal | |
Unconjugated | |
FAM13C | |
1200015N20Rik; C030038O19Rik; Fam13c; FAM13C1; family with sequence similarity 13 member C; family with sequence similarity 13, member C; family with sequence similarity 13, member C1; mKIAA1796; protein FAM13C; RGD1310149 | |
Rabbit | |
Affinity chromatography | |
RUO | |
220965 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NE31 | |
FAM13C | |
Synthetic peptide directed towards the N-terminal of human FAM13C1 (aa 110-159). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction