Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM148B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561364
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561364 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561364 Supplier Invitrogen™ Supplier No. PA561364
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PCTLESPIRA AAVPRRCAAE SDL Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 61%.

FAM148B (or C2CD4B) may be involved in inflammatory process. The protein may regulate cell architecture and adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM148B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C2CD4B
Gene Accession No. A6NLJ0
Gene Alias C2 calcium dependent domain containing 4B; C2 calcium-dependent domain containing 4B; C2 calcium-dependent domain-containing protein 4B; C2CD4B; FAM148B; family with sequence similarity 148, member B; NLF2; nuclear localized factor 2; nuclear-localized factor 2; Protein FAM148B
Gene Symbols C2CD4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM148B. Recombinant protein control fragment (Product #RP-100779).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388125
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.