Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM149A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563048 Supplier Invitrogen™ Supplier No. PA563048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGLPPVSPRD CVKDAVAAEV FDHVWTNMVE LLEELIRKHW ETTLTEGKKQ RETLKVAGNR FPHVLVPHAH ADGASGPPSG HAEAHGISLA SRLNPPQIHH FSSSF Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 58%.

FAM149A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM149A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM149A
Gene Accession No. A5PLN7
Gene Alias DKFZP564J102; FAM149A; family with sequence similarity 149 member A; family with sequence similarity 149, member A; MST119; MSTP119; Protein FAM149A
Gene Symbols FAM149A
Host Species Rabbit
Immunogen Recombinant protein fragment of Human FAM149A (Product #RP-100101)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.