Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM149B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558687 Supplier Invitrogen™ Supplier No. PA558687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERDSTIFGIR GKKLHFSSSY AHKASSIAKS SSFCSMERDE EDSIIVSEGI IEEYLAFDHI DIEEG Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 71%.

Involved in cilium assembly and protein localization to cilium. Predicted to be located in cilium. Implicated in Joubert syndrome. FAM149B1 (Family With Sequence Similarity 149 Member B1) is a Protein Coding gene. Diseases associated with FAM149B1 include Joubert Syndrome 36 and Orofaciodigital Syndrome Vi. An important paralog of this gene is FAM149A. Involved in the localization of proteins to the cilium and cilium assembly. Indirectly regulates the signaling functions of the cilium, being required for normal SHH/smoothened signaling and proper development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM149B1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM149B1
Gene Accession No. Q96BN6
Gene Alias FAM149B1; family with sequence similarity 149 member B1; family with sequence similarity 149, member B1; KIAA0974; Protein FAM149B1; RGD1359592
Gene Symbols FAM149B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM149B1. Recombinant protein control fragment (Product #RP-97087).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.