Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM14A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569661
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569661 Supplier Invitrogen™ Supplier No. PA569661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 77%; Human: 100%.

IFI27L2 plays a role in the apoptotic process and has a pro-apoptotic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM14A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IFI27L2
Gene Accession No. Q9H2X8
Gene Alias FAM14A; family with sequence similarity 14, member A; IFI27L2; interferon alpha inducible protein 27 like 2; interferon alpha-inducible protein 27-like protein 2; interferon, alpha-inducible protein 27-like 2; interferon-stimulated gene 12b protein; ISG12(b); ISG12(b) protein; ISG12B; pIFI27-like protein; Protein TLH29; TLH29; Unknown (protein for MGC:140204)
Gene Symbols IFI27L2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FAM14A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83982
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.