Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM151A Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297555V
Description
FAM151A Polyclonal specifically detects FAM151A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FAM151A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C1orf179, chromosome 1 open reading frame 179, family with sequence similarity 151, member A, hyporthetical protein MGC27169, hypothetical protein LOC338094, MGC27169 | |
Produced in rabbits immunized with E. coli-derived Human FAM151A fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
338094 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction