Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM151A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553502 Supplier Invitrogen™ Supplier No. PA553502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTANETGVPI MAHPPTIYSD NTLEQWLDAV LGSSQKGIKL DFKNIKAVGP SLDLLRQLTE EGKVRRPIWI NADILKGPNM LISTEVNATQ FLALVQEKYP KATLSPGWTT FYMSTSPNRT YTQAMVEKMH ELVG Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 81%.

The function of this protein has not been specifically defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM151A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM151A
Gene Accession No. Q8WW52
Gene Alias 2010309H15Rik; C1orf179; f151; FAM151A; family with sequence similarity 151 member A; family with sequence similarity 151, member A; family with sequence simliarity 151, member A; hyporthetical protein MGC27169; protein FAM151A; UNQ3034/PRO9836
Gene Symbols FAM151A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM151A. Recombinant protein control fragment (Product #RP-90515).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338094
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.