Learn More
Invitrogen™ FAM151B Monoclonal Antibody (OTI1F5)

Description
FAM151B Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | FAM151B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1F5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FAM151B |
| Gene Accession No. | Q6UXP7 |
| Gene Alias | 4930405M20Rik; AASA9217; f151; FAM151B; family with sequence similarity 151 member B; family with sequence similarity 151, member B; protein FAM151B; UNQ9217; UNQ9217/PRO28685 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.