Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM151B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113013
Description
FAM151B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
FAM151B is a protein coding gene.
Specifications
| FAM151B | |
| Polyclonal | |
| Unconjugated | |
| FAM151B | |
| 4930405M20Rik; AASA9217; f151; FAM151B; family with sequence similarity 151 member B; family with sequence similarity 151, member B; protein FAM151B; UNQ9217; UNQ9217/PRO28685 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 167555, 73942 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.25 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6UXP7 | |
| FAM151B | |
| Recombinant Human Protein FAM151B protein (1-276AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction