Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM153B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563137 Supplier Invitrogen™ Supplier No. PA563137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DPDTLAELLI RDVLQELSSY NGEEEDPEEV KTSLGVPQ Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 39%.

FAM153B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM153B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM153B
Gene Accession No. P0C7A2
Gene Alias FAM153B; family with sequence similarity 153 member B; family with sequence similarity 153, member B; protein FAM153B
Gene Symbols FAM153B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM153B. Recombinant protein control fragment (Product #RP-104499).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202134
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.