Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM155A Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298011R
Description
FAM155A Polyclonal specifically detects FAM155A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
FAM155A | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
family with sequence similarity 155, member A | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
728215 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 550 | |
50mM Sodium Borate | |
NALF1 | |
Produced in rabbits immunized with E. coli-derived Human FAM155A fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction