Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM155A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558792 Supplier Invitrogen™ Supplier No. PA558792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNRGKDDRGK ALFLGNSAKP VWRLETCYPQ GASSGQCFTV ENADAVCARN WSRGAAGGDG QEVRSKHPTP L Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 28%.

FAM155A gene ontology annotations related to this gene include calcium ion import across plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM155A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM155A
Gene Accession No. B1AL88
Gene Alias 6430500D16; AW121567; Fam155a; family with sequence similarity 155 member A; family with sequence similarity 155, member A; RGD1559498; Tmem28; transmembrane protein 28; transmembrane protein FAM155A
Gene Symbols FAM155A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM155A. Recombinant protein control fragment (Product #RP-98014).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728215
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.