Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM155A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988377
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8377 Supplier Bioss Supplier No. BS8223R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM155A gene ontology annotations related to this gene include calcium ion import across plasma membrane.

Specifications

Antigen FAM155A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM155A
Gene Accession No. B1AL88
Gene Alias FAM155A; family with sequence similarity 155 member A; family with sequence similarity 155, member A; Transmembrane protein FAM155A
Gene Symbols FAM155A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM155A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728215
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.