Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM155A Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 249291AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-4623


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

FAM155A gene ontology annotations related to this gene include calcium ion import across plasma membrane.
Specifications

Specifications

FAM155A
Polyclonal
Unconjugated
FAM155A
FAM155A, Transmembrane protein FAM155A
Rabbit
Antigen Affinity Chromatography
RUO
728215
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
B1AL88
FAM155A
FAM155A Fusion Protein Ag21575
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.