Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM155B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557517 Supplier Invitrogen™ Supplier No. PA557517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFSSNQGGGG LGLETLPALE EGLTREE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 93%.

TRIM41 belongs to a family of tripartite motif (TRIM) proteins defined as containing a RING finger, one or more B-box domains, and a coiled-coil region (Tanaka et al., 2005 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM155B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM155B
Gene Accession No. O75949
Gene Alias bB57D9.1; bB57D9.1 (TED protein); CXorf63; FAM155B; family with sequence similarity 155 member B; family with sequence similarity 155, member B; Protein TED; TED; TMEM28; Tmem28l; Transmembrane protein 28; transmembrane protein 28-like; transmembrane protein FAM155B
Gene Symbols FAM155B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM155B. Recombinant protein control fragment (Product #RP-95443).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.