Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM155B Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30407420UL
Description
FAM155B Polyclonal specifically detects FAM155B in Mouse, Rat samples. It is validated for Western Blot.Specifications
FAM155B | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
27112 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
chromosome X open reading frame 63, CXorf63, family with sequence similarity 155, member B, Protein TED, TEDbB57D9.1 (TED protein), TMEM28, Transmembrane protein 28bB57D9.1, transmembrane protein FAM155B | |
A synthetic peptide of Human FAM155B. | |
20 μL | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction