Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM158A Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734162 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4162 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4162 Supplier Proteintech Group Inc Supplier No. 239191AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EMC9 (ER Membrane Protein Complex Subunit 9) is a Protein Coding gene. An important paralog of this gene is EMC8.

Specifications

Antigen FAM158A
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EMC9
Gene Accession No. Q9Y3B6
Gene Alias C14orf122, CGI 112, FAM158A, UPF0172 protein FAM158A
Gene Symbols EMC9
Host Species Rabbit
Immunogen FAM158A Fusion Protein Ag20744
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51016
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.