Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM160A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561221 Supplier Invitrogen™ Supplier No. PA561221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLHHKPILKP LMMLLSSCSG TTTPTVEEKL VVLLNQLCSI LAKDPSILEL FFHTSEDQGA ANFLIFS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM160A1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM160A1
Gene Accession No. Q05DH4
Gene Alias FAM160A1; family with sequence similarity 160 member A1; family with sequence similarity 160, member A1; protein FAM160A1
Gene Symbols FAM160A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM160A1. Recombinant protein control fragment (Product #RP-100197).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729830
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.