Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM160A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563230
Description
Immunogen sequence: GSRTKKRSLL PEEDRNNVGE GEEEELGRRG RAGGAGEGPG HLPPPQLNGV PGSWPEGAKK VRLVPKEGAG ELL Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 32%.
FAM160A2 is a component of the FTS/Hook/FHIP complex (FHF complex). The FHF complex may function to promote vesicle trafficking and/or fusion via the homotypic vesicular protein sorting complex (the HOPS complex).
Specifications
FAM160A2 | |
Polyclonal | |
Unconjugated | |
FAM160A2 | |
C11orf56; FAM160A2; family with sequence similarity 160 member A2; family with sequence similarity 160, member A2; FHIP; FTS and Hook interacting protein; FTS and Hook-interacting protein; KIAA1759; Protein FAM160A2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84067 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N612 | |
FAM160A2 | |
Recombinant protein corresponding to Human FAM160A2. Recombinant protein control fragment (Product #RP-101644). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction