Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM160A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566651
Description
Immunogen sequence: ARQPLLRHGPV REALLTLLDA CGRPVPSSPA LDEGLVLLLS QLCVCVAQEP SLLEFFLQPP PEPGAAPRLL LFS Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
FAM160A2 is a component of the FTS/Hook/FHIP complex (FHF complex). The FHF complex may function to promote vesicle trafficking and/or fusion via the homotypic vesicular protein sorting complex (the HOPS complex).
Specifications
FAM160A2 | |
Polyclonal | |
Unconjugated | |
FAM160A2 | |
C11orf56; FAM160A2; family with sequence similarity 160 member A2; family with sequence similarity 160, member A2; FHIP; FTS and Hook interacting protein; FTS and Hook-interacting protein; KIAA1759; Protein FAM160A2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84067 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N612 | |
FAM160A2 | |
Recombinant Human FAM160A2. Recombinant protein control fragment (Product #RP-107302). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction