Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM160B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570323
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
FAM160B1 is a protein coding gene.
Specifications
FAM160B1 | |
Polyclonal | |
Unconjugated | |
FAM160B1 | |
AI450540; bA106M7.3; Fam160b1; family with sequence similarity 160 member B1; family with sequence similarity 160, member B1; Kiaa1600; mKIAA1600; Protein FAM160B1; RGD1306116; zgc:162264 | |
Rabbit | |
Affinity chromatography | |
RUO | |
57700 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5W0V3 | |
FAM160B1 | |
synthetic peptide directed towards the N terminal region of human FAM160B1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction