Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM161A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556935 Supplier Invitrogen™ Supplier No. PA556935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSPKLLTVCK PFDLHASPHA SIKREKILAD IEADEENLKE TRWPYLSPRR KSPVRCAGVN PVPCNCNPPV PTVSSRGREQ AVRKSEKERM R Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 53%.

This gene belongs to the FAM161 family. It is expressed mainly in the retina. Mouse studies suggested that this gene is involved in development of retinal progenitors during embryogenesis, and that its activity is restricted to mature photoreceptors after birth. Mutations in this gene cause autosomal recessive retinitis pigmentosa-28. Alternatively spliced transcript variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM161A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM161A
Gene Accession No. Q3B820
Gene Alias 4930430E16Rik; FAM161 centrosomal protein A; FAM161A; FAM161A, centrosomal protein; FAM161Afl; FAM161Ash; family with sequence similarity 161 member A; family with sequence similarity 161, member A; protein FAM161A; Protein FAM161A-like protein; retinitis pigmentosa 28 (autosomal recessive); RGD1304999; RP28
Gene Symbols FAM161A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM161A. Recombinant protein control fragment (Product #RP-95005).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.