Learn More
Invitrogen™ FAM161A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114156
Description
Immunogen sequence: RKEWVPTITV PEPFQMMIRE QKKKEESMKS KSDIEMVHKA LKKQEEDPEY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Pig: 93%; Rabbit: 86%; Rat: 93%.
This gene belongs to the FAM161 family. It is expressed mainly in the retina. Mouse studies suggested that this gene is involved in development of retinal progenitors during embryogenesis, and that its activity is restricted to mature photoreceptors after birth. Mutations in this gene cause autosomal recessive retinitis pigmentosa-28. Alternatively spliced transcript variants have been identified.
Specifications
| FAM161A | |
| Polyclonal | |
| Unconjugated | |
| FAM161A | |
| 4930430E16Rik; FAM161 centrosomal protein A; FAM161A; FAM161A, centrosomal protein; FAM161Afl; FAM161Ash; family with sequence similarity 161 member A; family with sequence similarity 161, member A; protein FAM161A; Protein FAM161A-like protein; retinitis pigmentosa 28 (autosomal recessive); RGD1304999; RP28 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84140 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3B820 | |
| FAM161A | |
| Immunogen sequence: RKEWVPTITV PEPFQMMIRE QKKKEESMKS KSDIEMVHKA LKKQEEDPEY. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.