Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM161B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553931 Supplier Invitrogen™ Supplier No. PA553931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSRQIFPPES FADTEAGEEL SGDGLVLPRA SKLDEFLSPE EEIDSTSDST GSIYQNLQEL KQKGRWCLLE SLFQSDPESD ENLSEDEEDL ESFFQDKDRG MVQVQCPQAL RCGSTRRCSS LNNLPSNIPR PQTQPPS Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 67%.

FAM161B is a 647 amino acid protein that belongs to the FAM161 family. The gene that encodes FAM161B consists of approximately 16,413 bases and maps to human chromosome 14q24.3. Housing over 700 genes, chromosome 14 comprises nearly 3.5% of the human genome. Chromosome 14 encodes the Presenilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease. The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder 1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction. An inversion of the long arm of chromosome 14 is thought to be involved in T-cell chronic lymphocytic leukemia (CLL), a cancer that affects T lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM161B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM161B
Gene Accession No. Q96MY7
Gene Alias 9330169D17; 9830169C18Rik; c14_5547; C14orf44; Fam161b; family with sequence similarity 161 member B; family with sequence similarity 161, member B; protein FAM161B
Gene Symbols FAM161B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM161B. Recombinant protein control fragment (Product #RP-88870).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145483
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.