Learn More
Invitrogen™ FAM162A Polyclonal Antibody

Description
Immunogen sequence: EGKKAAQRHET LTSLNLEKKA RLKEEAAMKA K Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.
Specifications
Specifications
| Antigen | FAM162A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FAM162A |
| Gene Accession No. | Q96A26 |
| Gene Alias | 2310056P07Rik; C3orf28; DC16; E2IG5; E2-induced gene 5 protein; E2-induced gene 5 protein homolog; f162; Fam162a; family with sequence similarity 162 member A; family with sequence similarity 162, member A; FWP001; Growth and transformation-dependent protein; HGTD-P; HIF-1 alpha-responsive proapoptotic molecule; protein FAM162A |
| Gene Symbols | FAM162A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.