Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM164A Monoclonal Antibody (OTI4D4), TrueMAB™, OriGene
SDP

Catalog No. 502539867
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9867 30 μL
50-167-5709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9867 Supplier OriGene Supplier No. TA505174S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAM164A Monoclonal antibody specifically detects FAM164A in Human samples. It is validated for Western Blot

Specifications

Antigen FAM164A
Applications Western Blot
Classification Monoclonal
Clone OTI4D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ALX4
Gene Accession No. Q9H161
Gene Alias 3110050N22Rik; AI790358; AU023959; C14H8ORF70; C8orf70; CGI-62; Fam164a; family with sequence similarity 164, member A; protein FAM164A; RGD1311970; UPF0418 protein FAM164A; Zc2hc1a; Zinc finger C2HC domain-containing protein 1A; zinc finger C2HC-type containing 1A; zinc finger, C2HC-type containing 1A
Host Species Mouse
Immunogen Full length human recombinant protein of human ALX4 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60529
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.