Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM164B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988381
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8381 Supplier Bioss Supplier No. BS8227R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZC2HC1B gene ontology annotations related to this gene include metal ion binding.

Specifications

Antigen FAM164B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZC2HC1B
Gene Accession No. Q5TFG8
Gene Alias BOS_9889; C1H6orf94; C6orf94; C9H6orf94; dJ468K18.5; FAM164B; family with sequence similarity 164, member B; protein FAM164B; ZC2HC1B; Zinc finger C2HC domain-containing protein 1B; zinc finger C2HC-type containing 1B; zinc finger, C2HC-type containing 1B
Gene Symbols ZC2HC1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM164B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153918
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.