Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM166A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571103 Supplier Thermo Scientific Supplier No. PA571103
Add to Cart
Add to Cart

FAM166A Polyclonal Antibody

FAM166A Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM166A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM166A
Gene Accession No. Q6J272
Gene Alias 4931415M17; 4931415M17 mRNA; Fam166a; family with sequence similarity 166 member A; family with sequence similarity 166, member A; HSD46; Protein FAM166A
Gene Symbols FAM166A
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human FAM166A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401565
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.