Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM167B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566455
Description
Immunogen sequence: PSYLEWTAQVQ SQAWRRAQAK PGPGGPGDIC GFDSMDSALE WLRRELREM Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.
FAM167B is a protein coding gene.
Specifications
FAM167B | |
Polyclonal | |
Unconjugated | |
FAM167B | |
C1orf90; FAM167B; family with sequence similarity 167 member B; family with sequence similarity 167, member B; protein FAM167B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84734 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BTA0 | |
FAM167B | |
Recombinant Human FAM167B. Recombinant protein control fragment (Product #RP-107128). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction