Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM168B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549398
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549398 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549398 Supplier Invitrogen™ Supplier No. PA549398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with bovine and rat based on sequence homology.

Inhibitor of neuronal axonal outgrowth. Acts as a negative regulator of CDC42 and STAT3 and a positive regulator of STMN2. Positive regulator of CDC27. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM168B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FAM168B
Gene Accession No. A1KXE4, Q80XQ8
Gene Alias Fam168b; family with sequence similarity 168 member B; family with sequence similarity 168, member B; KIAA0280L; Kiaa4042; MANI; mKIAA4042; myelin associated neurite outgrowth inhibitor; myelin-associated neurite-outgrowth inhibitor; p20; protein FAM168B
Gene Symbols FAM168B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 165-199 amino acids from the C-terminal region of human FAM168B.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130074, 214469
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.