Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM168B Polyclonal Antibody
FAM168B Polyclonal Antibody
Supplier: Thermo Scientific PA563977
Description
FAM168B Polyclonal Antibody for IHC

Specifications
FAM168B | |
Polyclonal | |
Unconjugated | |
FAM168B | |
Fam168b; family with sequence similarity 168 member B; family with sequence similarity 168, member B; KIAA0280L; Kiaa4042; MANI; mKIAA4042; myelin associated neurite outgrowth inhibitor; myelin-associated neurite-outgrowth inhibitor; p20; protein FAM168B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
130074 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A1KXE4 | |
FAM168B | |
Recombinant protein corresponding to Human FAM168B. Recombinant protein control fragment (Product #RP-103811). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction