Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM168B Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 244221AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-4350


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Modulates neuronal axonal outgrowth by acting as a negative regulator of CDC42 and STAT3 and a positive regulator of STMN2. Positive regulator of CDC27.
Specifications

Specifications

FAM168B
Polyclonal
Unconjugated
FAM168B
FAM168B, KIAA0280L, p20, Protein FAM168B
Rabbit
Antigen Affinity Chromatography
RUO
130074
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.44 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A1KXE4
FAM168B
FAM168B Fusion Protein Ag19535
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.