Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM169A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA559630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559630 Supplier Thermo Scientific Supplier No. PA559630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPFNSSEDST NLVPLVVESS KPPEVDAPDK TPRIPDSEML MDEGTSDEKG HMEEKLSLLP RKKAHLGSSD NVATMSNEER SDGGFPNSVI AEFSEEP Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 59%.

FAM169A Polyclonal antibody specifically detects FAM169A in Human samples. It is validated for Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM169A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM169A
Gene Accession No. Q9Y6X4
Gene Alias FAM169A; family with sequence similarity 169 member A; family with sequence similarity 169, member A; KIAA0888; Protein FAM169A; SLAP75; soluble lamina-associated protein of 75 kD; Soluble lamin-associated protein of 75 kDa
Gene Symbols FAM169A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM169A
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26049
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.