Learn More
FAM170A Mouse anti-Human, Clone: OTI6E4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809624

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Acts as a nuclear transcription factor that positively regulates the expression of heat shock genes. Binds to heat shock promoter elements (HSE).Specifications
FAM170A | |
Monoclonal | |
Unconjugated | |
FAM170A | |
ZNFD | |
Mouse | |
Affinity Chromatography | |
RUO | |
340069 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6E4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
A1A519 | |
FAM170A | |
Full length human recombit protein of human FAM170A produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.